(3,5-Dichlorophenoxy)acetonitrile 97%
Jump to:
About this item
Purity: 97% CAS No.: 103140-12-1 Molecular Formula: C8H5Cl2NO Formula Weight: 202.04 Melting Point: 58-60° Beilstein Reference: 4983677 UN No.: 3439 MDL No.: MFCD00052707
- CAS Number:103140-12-1
- Formula:C₈H₅Cl₂NO
- MDL Number:MFCD00052707
- Molecular Weight:202.04 g/mol
- Melting Pt:58…60 °C
- Storage Temperature:Ambient
- UN:3439
- ADR:6.1,III
- ADR:"6.1",III
0 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 103140-12-1 Molecular Formula: C8H5Cl2NO Formula Weight: 202.04 Melting Point: 58-60° Beilstein Reference: 4983677 UN No.: 3439 MDL No.: MFCD00052707
- CAS Number:103140-12-1
- Formula:C₈H₅Cl₂NO
- MDL Number:MFCD00052707
- Molecular Weight:202.04 g/mol
- Melting Pt:58…60 °C
- Storage Temperature:Ambient
- UN:3439
- ADR:6.1,III
- ADR:"6.1",III
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Product Family Options
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



