Anti-Streptavidin Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemical
Certificates
About this item
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:3000 - 1:12000; Immunochemistry: 1:1500 - 1:5000;
Type: Tag
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptomyces avidinii
Specifications
- Catalog No:
- RL200-401-095S
- RL200-401-095
- Antigen Symbol:
- Streptavidin
- Streptavidin
- Antigen Name:
- Streptavidin
- Streptavidin
- Antibody Type:
- Tag
- Tag
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- S. avidinii
- S. avidinii
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Streptavidin (Streptomyces avidinii)
- Streptavidin (Streptomyces avidinii)
- Format:
- Antibody fraction
- Antibody fraction
- Form:
- Liquid (sterile filtered)
- Liquid (sterile filtered)
- Purification:
- multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- Concentration:
- 1.0 mg/ml by UV absorbance at 280 nm
- 1.0 mg/ml by UV absorbance at 280 nm
- Cross Adsorption:
- No
- No
- Storage Buffer:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:
- –20 °C
- –20 °C
- Shipping Temperature:
- Dry Ice
- Dry Ice
Specifications
Related Information
Frequently Bought Together