Anti-RFP Rabbit Polyclonal Antibody
Catalog # RL600-401-379S
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Tag
- Antigen Name:Red Fluorescent Protein
- Antigen Symbol:RFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:25 µL
- Epitope:Full length
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen Synonyms:DSRED
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:The immunogen is a Red Fluorescent Protein (RFP) fusion Protein corresponding to the full length amino acid sequence (234aa) derived from the mushroom polyp coral Discosoma.
- Purification:affinity-purified
- Cat. No.:RL600-401-379S
- Supplier no.:600-401-379S
Specifications
About this item
Polyclonal anti-RFP is designed to detect RFP and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen.
Recommended Dilutions: ELISA: 1:50000 - 1:100000; Immunofluorescence: 1:200-1:2000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:200-1:2000;
Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: 0