About this item
PE anti-human TCR Vα24-Jα18 (iNKT cell) [6B11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactivity: Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.
Type: Primary
Antigen: TCR Va24-Ja18
Clonality: Monoclonal
Clone: 6B11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-reactivity: Pigtailed Macaque;Sooty Mangabey
Specifications
- Catalog No:
- 342904-BL
- 342903-BL
- Antigen Symbol:
- TCR Va24-Ja18
- TCR Va24-Ja18
- Antigen Synonyms:
- TCR Vα24-JαQ|iNKT|invariant NKT cell|TCR Vα24-Jα18
- TCR Vα24-JαQ|iNKT|invariant NKT cell|TCR Vα24-Jα18
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 6B11
- 6B11
- Reactivity:
- Sooty mangabey, Pigtailed macaque, Human
- Sooty mangabey, Pigtailed macaque, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 28659
- 28659
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT