Anti-MACC1 Rabbit Polyclonal Antibody
RL600-401-C07
- Antibody Type:Primary
- Antigen Name:Metastasis Associated in Colon Cancer 1
- Antigen Symbol:MACC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:Center
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:346389
- Antigen Synonyms:7A5|MACC1|SH3BP4L|SH3 domain-containing protein 7a5|Metastasis-associated in colon cancer protein 1
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:MACC1 Antibody was produced from whole rabbit serum prepared by repeated immunizations with a peptide corresponding to an internal region of human MACC1.
- Purification:affinity-purified
- Cat. No.:RL600-401-C07
- Supplier no.:600-401-C07
Anti-MACC1 antibody has been tested in ELISA, WB, and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~91-96 kDa in size corresponding to MACC1 by Western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1.0 - 2.0 µg/ml; Immunochemistry: 2.5 µg/ml;
Type: Primary
Antigen: MACC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human