Anti-IL34 Rabbit Polyclonal Antibody
Catalog # RL600-401-C06
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 34
- Antigen Symbol:IL34
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:146433
- Antigen Synonyms:C16orf77|Interleukin-34|IL-34
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Anti-IL-34 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminus of human IL-34 Protein.
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL600-401-C06
- Supplier no.:600-401-C06
Specifications
About this item
Anti-IL-34 Antibody has been tested for use in ELISA and WB. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~48 kDa in size corresponding to IL-34 by Western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 0.25 - 0.5 µg/ml;
Type: Primary
Antigen: IL34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human