Anti-D1MIT384 Rabbit Polyclonal Antibody
100 µL
Catalog # RL600-401-D45
Supplier: Rockland Immunochemical
: 600-401-D45
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Anti-GABA(A) Receptor alpha 2 (Rabbit) Antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 51 kDa in size corresponding to the alpha 2 subunit of the GABA(A) Receptor.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: D1MIT384
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Specifications
- Size:100 µL
- Cross Adsorption:No
- Clonality:Polyclonal
- Purification:affinity-purified
- Form:Liquid
- Storage Buffer:0.01 M HEPES, 0.15 M Sodium Chloride, pH 7.50.1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Isotype:IgG
- Host:Rabbit
- Cat. No.:RL600-401-D45
- Reactivity:Rat|Mouse
- Western Blot:Yes
- Format:Antibody fraction
- Shipping Temperature:Dry Ice
- Antibody Type:Primary
- Antigen Symbol:D1MIT384
- Gene ID:61856
- Immunogen:Anti-GABA(A) Receptor alpha 2 Antibody was produced in rabbit by repeated immunizations with a fusion Protein from the cytoplasmic loop of the rat receptor alpha 2 subunit.
- Conjugation:Unconjugated
- Antigen Name:DNA segment, Chr 1, Massachusetts Institute of Technology 384
- Storage Temperature:–20 °C



