Anti-DNM1 Rabbit Polyclonal Antibody
Catalog # RL600-401-D31
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Name:Dynamin 1
- Antigen Symbol:DNM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µL
- Epitope:N-Terminal
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:1759
- Antigen Synonyms:EC 3.6.5.5|DNM1|Dynamin BREDNM19|DNM|brain|dynamin-1|Dynamin|B-dynamin|D100
- Storage Buffer:0.01 M HEPES, 0.15 M Sodium Chloride, pH 7.50.1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage Temperature:–20 °C
- Shipping Temperature:Dry Ice
- Immunogen:Anti-Dynamin Antibody was produced in rabbit by repeated immunizations with synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH.
- Purification:affinity-purified
- Cat. No.:RL600-401-D31
- Supplier no.:600-401-D31
Specifications
About this item
Anti-Dynamin Antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 95 kDa in size corresponding to the dynamin Proteins in the appropriate cell lysate.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000
Type: Primary
Antigen: DNM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat