Anti-IgG (minimal x-reactivity) Donkey Polyclonal Antibody (D488) [clone: Poly4064]
Catalog # 406404-BL
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:IgG (minimal x-reactivity)
- Clonality:Polyclonal
- Clone:Poly4064
- Conjugation:DyLight® 488
- Flow Cytometry:Yes
- Host:Donkey
- ImmunoFluorescence:Yes
- Reactivity:Rabbit
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Gene ID:100343867
- Antigen Synonyms:IgG
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography, and conjugated with DyLight™ 488 under optimal conditions.
- Cat. No.:406404-BL
- Supplier no.:406404
Specifications
About this item
DyLight™ 488 Donkey anti-rabbit IgG (minimal x-reactivity) [Poly4064]; Isotype: Donkey Polyclonal Ig; Reactivity: Rabbit; Apps: FC, IF; Size: 100 μg
It is recommended that the reagent be titrated for optimal performance for each application.
* DyLight™ 488 has a maximum absorption of 493nm and a maximum emission of 518 nm (similar to Alexa Fluor® 488 or FITC).
Type: Primary
Antigen: IgG (minimal x-reactivity)
Clonality: Polyclonal
Clone: Poly4064
Conjugation: D488
Epitope:
Host: Donkey
Isotype: Donkey Polyclonal Ig
Reactivity: Rabbit