To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Delta-like 4
- Antigen Symbol:DLL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:25 µL
- Environmentally Preferable:
- Epitope:Center
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:54567
- Antigen Synonyms:hdelta2|DLL4
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.0mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Anti-DELTA-4 antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of Human DELTA-4.
- Purification:affinity-purified
- Cat. No.:RL600-401-696S
- Supplier no.:600-401-696S
Specifications
About this item
Anti-DELTA-4 has been tested for use in ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 74 kDa in size corresponding to DELTA-4 by western blotting.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:2500; Immunochemistry: 10 µg/ml to 20 µg/ml;
Type: Primary
Antigen: DLL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Partial Reactivity To Mouse And Rat