ContactUsLinkComponent
Alkaline Phosphatase Enhancer, Diagnostic BioSystems
Alkaline Phosphatase Enhancer, Diagnostic BioSystems
Catalog # 99990-072
CAS Number:  
Alkaline Phosphatase Enhancer, Diagnostic BioSystems
Catalog # 99990-072
Supplier Number:  K011
CAS Number:  
Specifications
  • Description:
    Alkaline Phosphatase Enhancer
  • Size:
    250 ml
  • Cat. No.:
    99990-072
  • Supplier no.:
    K 011
Specifications
About this item
Alkaline phosphatase enhancer is a ready to use stable solution designed to increase the signal generated by alkaline phosphatase several fold. A single treatment of tissue sections with enhancer before the treatment with substrate/chromogen enables the visualization of difficult to localize antigens.
Alkaline phosphatase, an enzyme derived from bovine intestinal mucosa, is often used as a label for in situ hybridization and immunohistochemistry.
Specimens stained with the substrates/chromogens cannot be dehydrated in ethanol and should be mounted in aqueous based mounting medium such as C/C mount.