SP Bel-Art Spinbar® Stir Bar Restrainer, Bel-Art Products, a part of SP
Catalog # 89042-240
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Restrainer spinbar
- Cat. No.:89042-240
- Supplier no.:377861000
Specifications
About this item
The Restrainer fits ergonomically in one hand, and when placed alongside the outer wall of a mixing vessel it retains and secures the stirring bar in situ.
- Ergonomic design
- Confined stirring bar can be retrieved without contaminating solutoin
- Strong magnetic restrains magnetic stirring bars
After the vessel is drained, the confined stirring bar is securely retrieved from the vacant container without contaminating the solution. It houses a strong magnet that restrains magnetic stirring bars up to 80 mm (3") long.