Venusil XBP C18 HPLC Columns, Bonna-Agela Technologies
Supplier: Phenomenex
Certificates
About this item
Venusil XBP C18 columns offer the maximum bonding density, and therefore provide the highest hydrophobicity (the lowest polarity).
- Broad pH range (1.5 to 10.0 )
- Low pH, 0.2% TFA, high pH, 0.010 M triethylamine
- Surface: 380m²/g
- Pore Size: 100Å
Columns offer unmatched inertness, resulting in one of the most base-friendly columns available. These highly efficient columns are excellent at any applicable pH. They enable large volume injection, and maintain very high efficiency with very large injection volumes.
This allows for the least interaction between the analytes and the silanol groups. Venusil XBP columns have extraordinary column stability at high pHs.
Frequently Bought Together