Anti-GPCR TM7XN1/GPR56 Rabbit Polyclonal Antibody
Catalog # 89366-448
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Adhesion G Protein-Coupled Receptor G1
- Antigen Symbol:ADGRG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25 μg
- Gene ID:14766
- Antigen Synonyms:BFPP|GPR56|BPPR|TM7XN1|TM7LN4
- Storage Buffer:Phosphate buffered saline, pH 7.7, containing 0.01% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide - KLH conjugated. Peptide sequence identity/similarity to Mouse: 75/87%. Rat: 68/87%.
- Tested Applications:IHC-P
- Purification:Peptide immunogen affinity column
- Cat. No.:89366-448
- Supplier no.:GTX77957
Specifications
About this item
Rabbit polyclonal to G Protein-Coupled Receptor TM7XN1/GPR56
Type: Primary
Antigen: ADGRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human