To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Placental Alkaline Phosphatase
- Antigen Symbol:PLAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:500 μl
- Environmentally Preferable:
- Gene ID:250
- Antigen Synonyms:placental alkaline phosphatase 1|Regan isozyme|PALP|alkaline phosphatase Regan isozyme|alkaline phosphatase|ALP|alkaline phosphomonoesterase|PLAP|PLAP-1|glycerophosphatase|placental type
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing 0.2% BSA and 0.09% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:Purified PLAP from human placenta
- Tested Applications:IHC-P, IHC-Fr, IP, WB
- Purification:Protein A affinity purified
- Cat. No.:89353-088
- Supplier no.:GTX72989
Specifications
About this item
Rabbit polyclonal to Placental Alkaline Phosphatase
Immunoblot 1:100-1:500. Immunoprecipitation, Immunohistochemistry 1:50. The optimal conditions should be determined by the individual laboratory. This antibody can be used on formalin-fixed, paraffin-embedded tissue sections. Prolonged fixation in buffered formalin can destroy the epitope. The antibody may be used at a dilution of 1:50 with AutoProbe III (GTX28620). It is recommended that this product be used on frozen tissue sections or specimens. The optimal conditions should be determined by the individual laboratory.
Type: Primary
Antigen: PLAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human