Anti-HEC1 Mouse Monoclonal Antibody [clone: 9G3.23]
Catalog # 89364-838
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Ndc80 Kinetochore Complex Component
- Antigen Symbol:HEC1
- Clonality:Monoclonal
- Clone:9G3.23
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10403
- Antigen Synonyms:HsHec1|KNTC2|TID3|hsNDC80|HEC|HEC1
- Amino Acid Number:56 to 642
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing no preservatives
- Molecular Weight:80 kDa
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.27 mg/ml
- Immunogen:Human HEC1 protein consisting of amino acids 56-642.
- Tested Applications:ICC/IF, IP, WB
- Purification:Protein G Affinity Purified
- Cat. No.:89364-838
- Supplier no.:GTX70268
Specifications
About this item
Mouse monoclonal antibody to human HEC1
For ICC/IF: Use at a concentration of 4-10 µg/ml. For IP: Use at a concentration of 1-5 µg/ml. For WB: Use at a dilution of 1:500-1:3000.For FACS:Assay dependent Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: HEC1
Clonality: Monoclonal
Clone: 9G3.23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human