Anti-JAB1 Mouse Monoclonal Antibody [clone: 2A10]
89364-764
: Genetex
- Antibody Type:Primary
- Antigen Name:Cop9 Signalosome Subunit 5
- Antigen Symbol:JAB1
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10987
- Antigen Synonyms:JAB1|CSN5|SGN5|MOV-34
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing no preservatives
- Molecular Weight:38 kDa
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:The full-length mouse JAB-1 gene expressed in E. coli.
- Tested Applications:ICC/IF, IHC, IP, WB
- Purification:Protein G Affinity Purified
- Cat. No.:89364-764
- Supplier no.:GTX70203
Mouse monoclonal antibody to human Jab1
For IP: Use at a concentration of 0.5 - 1 µg/ml. For WB: Use at a dilution of 1:500-1:3000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: JAB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human