To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:E2F Transcription Factor 1
- Antigen Symbol:E2F1
- Clonality:Monoclonal
- Clone:16G7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:1869
- Antigen Synonyms:Retinoblastoma binding protein 3|RBBP-3|PRB-binding protein E2F-1|Retinoblastoma-associated protein 1|RBAP-1|E2F-1|Transcription factor E2F1|E2F1|PBR3|RBBP3
- Amino Acid Number:244 to 264
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing no preservatives
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:The region encoding the DNA binding domain (amino acids 244-264) of E2F-1 expressed in E. coli.
- Tested Applications:ICC/IF, IHC-P, IP, WB
- Purification:Protein G Affinity Purified
- Cat. No.:89364-726
- Supplier no.:GTX70163
Specifications
About this item
Mouse monoclonal [16G7] antibody to E2F - 1
For ICC/IF: 1:100-1:1000. For IP: Use at an assay dependent dilution. For WB: 1:500-1:3000. For IHC-P: 1:100-1:1000. Predicted molecular weight: 47 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: E2F1
Clonality: Monoclonal
Clone: 16G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human