Order Entry
United States
ContactUsLinkComponent
Anti-HEY1 Rabbit Polyclonal Antibody
Anti-HEY1 Rabbit Polyclonal Antibody
Catalog # 89363-506
Supplier:  Genetex
Anti-HEY1 Rabbit Polyclonal Antibody
Catalog # 89363-506
Supplier:  Genetex
Supplier Number:  GTX42614

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Hairy/enhancer-of-split related with YRPW motif 1
  • Antigen Symbol:
    HEY1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    23462
  • Antigen Synonyms:
    MGC1274|HESR1|BHLHb31|OAF1|HERP2|CHF2|HRT-1
  • Storage Buffer:
    Preservative: None; Constituents: 2% Sucrose, PBS
  • Storage Temperature:
    Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
  • Concentration:
    1.0 mg/ml
  • Immunogen:
    Synthetic peptide derived from residues 207-261 of Human HEY1.
  • Tested Applications:
    IHC-P
  • Purification:
    Protein A affinity purified
  • Cat. No.:
    89363-506
  • Supplier no.:
    GTX42614

Specifications

About this item

Rabbit polyclonal to HEY1

IHC-P: Use at a concentration of 4 - 8 ?g/ml. WB: Use at a concentration of 2.5 ?g/ml in 5% skim milk / PBS buffer, and HRP conjugated anti-Rabbit IgG should be diluted in 1:50,000 - 100,000 as second antibody. Predicted molecular weight: 34 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HEY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human