To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Erythropoietin
- Antigen Symbol:EPO
- Clonality:Monoclonal
- Clone:Epo2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:500 μg
- Gene ID:2056
- Antigen Synonyms:EP|MVCD2
- Storage Buffer:Preservative: 0.1% Sodium Azide. Constituents: PBS. pH 7.4
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. We recommend to parafilm vials during storage to prevent evaporation of buffer.
- Concentration:3.3 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Tested Applications:ELISA, WB
- Purification:Protein G purified
- Cat. No.:89352-828
- Supplier no.:GTX40375
Specifications
About this item
Mouse monoclonal [Epo2] to EPO
ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 21 kDa. Recommended pair for sandwich immunoassay:Capture: GTX40375; Detection: GTX40473. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: EPO
Clonality: Monoclonal
Clone: Epo2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human