To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tenascin C
- Antigen Symbol:TNC
- Clonality:Monoclonal
- Clone:BC-24
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Format:Ascites
- Gene ID:3371
- Antigen Synonyms:HXB|JI|TN|GMEM|150-225|GP|DFNA56|TN-C
- Storage Buffer:Ascitic fluid with 0.1% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Full length native protein (purified) (Human).
- Tested Applications:Dot Blot, Radio Immunoassay
- Purification:Ascites
- Cat. No.:89361-910
- Supplier no.:GTX26393
Specifications
About this item
Mouse monoclonal [BC - 24] to Tenascin
Dot: Use at an assay dependant dilution. ELISA: Use at an assay dependant dilution. IHC-F: Use at an assay dependant dilution. ICC: Use at an assay dependant dilution. IHC-P: Use at a dilution of 1/4000. Perform enzymatic antigen retrieval before commencing with IHC staining protocol. IHC-Fr: Use at an assay dependant dilution. RIA: Use at an assay dependant dilution. WB: Use at an assay dependant dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: TNC
Clonality: Monoclonal
Clone: BC-24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human