To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Microtubule Associated Protein Tau
- Antigen Symbol:MAPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μl
- Environmentally Preferable:
- Gene ID:4137
- Antigen Synonyms:PPP1R103|FTDP-17|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
- Modification:phospho-Ser422
- Storage Buffer:Preservative: 0.05% Sodium Azide; Constituents: PBS,50% glycerol, 1mg/ml BSA. pH 7.3
- Storage Temperature:Aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
- Concentration:0.25 mg/ml
- Immunogen:Synthetic peptide (Human) derived from a region of human tau that contains serine 422. The region surrounding serine 422 is also conserved in mouse and rat.
- Tested Applications:WB
- Purification:Immunogen affinity purified
- Cat. No.:89361-562
- Supplier no.:GTX24862
Specifications
About this item
Rabbit polyclonal to Tau ( phospho S422 )
WB: Use at a concentration of 0.1 - 1.0 ?g/ml. Predicted molecular weight: 62 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human