To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:beta Subunit CHOLERA TOXIN
- Antigen Symbol:beta Subunit CHOLERA TOXIN
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Storage Buffer:PBS, pH 7.2, sodium azide, 0.1%
- Storage Temperature:–20 °C
- Immunogen:Purified beta subunit of cholera toxin.
- Tested Applications:ELISA, WB
- Purification:purified by Protein A chromatography
- Cat. No.:89360-940
- Supplier no.:GTX21003
Specifications
About this item
Cholera toxin (CT) acts by transfer of ADP-ribose from intracellular NAD to the alpha chain of a trimeric G protein that functions to stimulate adenylate cyclase resulting in the increase of host cell cAMP. CT binds via the beta subunits to the GM1 ganglioside. Gangliosides are glycolipids that also contain oligosaccharides with one or more sialic acid residues.
Type: Primary
Antigen: beta SUBUNIT CHOLERA TOXIN
Clonality: Monoclonal
Clone: "ERROR:#N/A"
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: