Specifications
- Antibody Type:Primary
- Antigen Name:Serum Amyloid A
- Antigen Symbol:SAA1
- Clonality:Monoclonal
- Clone:115
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:6288
- Antigen Synonyms:SAA2|PIG4|SAA|TP53I4
- Storage Buffer:Phosphate-buffered saline, pH 7.2 containing no preservatives
- Molecular Weight:12 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Highly purified recombinant human serum amyloid A (MW: 12 kDa)
- Tested Applications:ELISA, WB
- Purification:Protein G affinity purified
- Cat. No.:89360-878
- Supplier no.:GTX20687
Specifications
About this item
The serum amyloid A (SAA) family comprises a number of differentially expressed lipoproteins, acute-phase SAA1 and SAA2, the former being a major component in plasma, and constitutive SAA's (C-SAAs). Although the liver is the primary site of synthesis of both SAA types, extrhepatic production has been reported. The in-vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed it's importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations. SAA rises earlier and more sharply than CRP.
For ELISA: Use at a concentration of 0.3 ?g/ml. For WB: Use at a concentration of 0.1-0.5 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: SAA1
Clonality: Monoclonal
Clone: 115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human