Anti-ATOH1 Rabbit Polyclonal Antibody
89356-906
: Genetex
- Antibody Type:Primary
- Antigen Name:atonal bHLH transcription factor 1
- Antigen Symbol:ATOH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:474
- Antigen Synonyms:bHLHa14|ATH1|HATH1|MATH-1
- Storage Buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:38 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MATH1. The exact sequence is proprietary.
- Tested Applications:ICC/IF, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89356-906
- Supplier no.:GTX111898
Rabbit Polyclonal antibody to MATH1 (protein atonal homolog 1)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: ATOH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human