Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Microtubule-associated protein 2
- Antigen Symbol:MAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Form:Lyophilized
- Gene ID:4133
- Antigen Synonyms:MAP2A|MAP2C|DKFZp686I2148|microtubule-associated protein 2|MAP-2|MAP2B
- Storage Buffer:Whole antisera
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:A synthetic peptide as a part of rat, mouse Mtap2 conjugated to an immunogenic carrier protein was used as the immunogen.
- Purification:Unpurified
- Cat. No.:102121-308
- Supplier no.:NB100-98717
Specifications
About this item
Sheep Polyclonal MAP2 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse, Rat.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:300-1:2000
Type: Primary
Antigen: MAP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse