Order Entry
United States
ContactUsLinkComponent
SD OLLAS/DYKDDDDK Vector, Novus Biologicals (NBA1-07085)
SD OLLAS/DYKDDDDK Vector, Novus Biologicals (NBA1-07085)
Catalog # 102136-986
Supplier:  Novus Biologicals
CAS Number:  
undefined
SD OLLAS/DYKDDDDK Vector, Novus Biologicals (NBA1-07085)
Catalog # 102136-986
Supplier:  Novus Biologicals
Supplier Number:  NBA1-07085
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 µg
  • Cat. No.:
    102136-986
  • Supplier no.:
    NBA1-07085

Specifications

About this item

The pCMV-NS-OLLAS vector provides a convenient vector to express recombinant proteins under control of the CMV promoter with both an N-terminal OLLAS tag and a DYKDDDDK tag. Using this vector the protein will be secreted from the cell due to the inclusion of a signal peptide. We recommend inserting your protein of interest into the vector in frame using the Xba I and EcoR I/Not I restriction sites shown above. Bacterial selection can be performed using Kanamycin, while Neomycin resistance can be used for screening mammalian transfectants. The OLLAS tag has been shown to be detectable at ~100-fold lower levels that similar commercially available tags using the OLLA-2 monoclonal antibody. The OLLAS tag has also been demonstrated to provide superior functionality whether used as an N-terminal, C-terminal, or internal tag on recombinant and fusion proteins without interfering with the biological activity of the tagged protein.

  • Buffer: 5 mM Tris pH 8.0
  • Specificity: pCMV-SD OLLAS DYKDDDDK Vector for the expression of secretory protein with N-terminally FLAG(DYKDDDDK) and OLLAS dual tandem tagged.
  • Tested applications: Western Blot

Application Notes: Using this vector the protein will be secreted from the cell due to the inclusion of a signal peptide. We recommend inserting your protein of interest into the vector in frame using the Xba I and EcoR I/Not I restriction sites shown above. Bacterial selection can be performed using Kanamycin, while Neomycin resistance can be used for screening mammalian transfectants.

Alternate Names for SD OLLAS/DYKDDDDK Vector:
DDDDK vector
DYKDDDDK epitope tag vector
DYKDDDDK vector
ECS tag vector
ECS vector
Enterokinase Cleavage Site epitope tag vector
Enterokinase Cleavage Site tag vector
FLAG vector (FLAG is a trademark of Sigma Aldrich)
OLLA-2 vector
xxxDDDDK epitope tag vector

Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.