Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:DDB1 and CUL4 associated Factor 13
- Antigen Symbol:WDSOF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:25879
- Antigen Synonyms:DDB1 and CUL4 associated factor 13|MGC138247|WD repeat and SOF domain-containing protein 1|WDSOF1|WD repeats and SOF1 domain containing|HSPC064|DKFZp564O0463|GM83|MGC126859
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to WDSOF1(WD repeats and SOF1 domain containing) The peptide sequence was selected from the N terminal of WDSOF1. Peptide sequence WSPAGRATEMKVKMLSRNPDNYVRETKLDLQRVPRNYDPALHPFEVPREY.
- Purification:Immunogen affinity purified
- Cat. No.:102200-092
- Supplier no.:NBP1-70747
Specifications
About this item
The WDSOF1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to WDSOF1. This antibody reacts with human. The WDSOF1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: WDSOF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human