Order Entry
United States
ContactUsLinkComponent
Anti-TIF1 alpha Rabbit Polyclonal Antibody
Anti-TIF1 alpha Rabbit Polyclonal Antibody
  102107-876
Anti-TIF1 alpha Rabbit Polyclonal Antibody
  102107-876
 :  NB100-2597

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    TIF1 alpha
  • Antigen Symbol:
    TIF1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    8805
  • Antigen Synonyms:
    EC 6.3.2.-|RNF82Tif1a|PTC6|TIF1ATIF1ALPHA|RING finger protein 82|E3 ubiquitin-protein ligase TRIM24|hTIF1|transcription intermediary factor 1-alpha|tripartite motif-containing 24|TIF1TF1A|Tripartite motif-containing protein 24|TIF1-alpha|transcriptional intermediary factor 1|EC 6.3.2|tripartite motif containing 24
  • Storage Buffer:
    Tris-Citrate/Phosphate (pH 7.0 - 8.0)
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    1.0 mg/ml
  • Immunogen:
    The immunogen recognized by this antibody maps to a region between residue 1,000 and the C-terminus (residue 1050) of human Transcriptional Intermediary Factor 1 Alpha (NP_056989.2).
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102107-876
  • Supplier no.:
    NB100-2597

 

 

The TIF1 alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to TIF1 alpha. This antibody reacts with human. The TIF1 alpha Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: TIF1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human