Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Synaptotagmin 17
- Antigen Symbol:SYT17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Lyophilized
- Gene ID:51760
- Antigen Synonyms:sytXVII|B/K protein|synaptotagmin-17|synaptotagmin XVIIProtein B/K
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:A synthetic peptide from the internal part of human Synaptotagmin 17 (SYT17) conjugated to an immunogenic carrier protein was used as the immunogen.
- Purification:IgG purified
- Cat. No.:102156-000
- Supplier no.:NBP1-03494
Specifications
About this item
The SYT17 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SYT17. This antibody reacts with human. The SYT17 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Type: Primary
Antigen: SYT17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human