Order Entry
United States
ContactUsLinkComponent
Anti-STMN2 Rabbit Polyclonal Antibody
Anti-STMN2 Rabbit Polyclonal Antibody
Catalog # 102179-042
Supplier:  Novus Biologicals
undefined
Anti-STMN2 Rabbit Polyclonal Antibody
Catalog # 102179-042
Supplier:  Novus Biologicals
Supplier Number:  NBP1-49461

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Stathmin 2
  • Antigen Symbol:
    STMN2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.05 ml
  • Gene ID:
    11075
  • Antigen Synonyms:
    SCGN10stathmin-2|neuron-specific growth-associated protein|FLJ34868|SCG10|stathmin-like 2|superior cervical ganglia|FLJ50995|Superior cervical ganglion-10 protein|Protein SCG10|neural specific 10|SCG10neuronal growth-associated protein (silencer element)
  • Storage Buffer:
    Whole antisera
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    C-terminal peptide of mouse STMN2. [Swiss-Prot P55821]
  • Tested Applications:
    In vivo assay
  • Purification:
    Unpurified
  • Cat. No.:
    102179-042
  • Supplier no.:
    NBP1-49461

Specifications

About this item

The Stathmin-2 / STMN2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Stathmin-2 / STMN2. This antibody reacts with human, mouse, rat, bovine. The Stathmin-2 / STMN2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen, In vivo assay.

Type: Primary
Antigen: STMN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine