Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Spectrin beta 3
- Antigen Symbol:SPTB3
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:6712
- Antigen Synonyms:Beta-III spectrin|GTRAP41|non-erythroid beta chain 2|SCA5|Spectrin|glutamate transporter EAAT4-associated protein 41|non-erythrocytic 2|spectrin beta chain|beta|spinocerebellar ataxia 5|brain 2|spectrin|KIAA0302
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The epitope recognized by this antibody maps to a region between residues 2150 and 2200 of Spectrin beta 3 using the numbering given in entry NP_008877.1
- Purification:Immunogen affinity purified
- Cat. No.:102123-718
- Supplier no.:NB110-58783G
The Spectrin beta 3 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to Spectrin beta 3. This antibody reacts with human. The Spectrin beta 3 Antibody [DyLight 488] has been validated for the following applications: Immunohistochemistry, Immunoprecipitation.
Type: Primary
Antigen: Spectrin beta 3
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human