Order Entry
United States
ContactUsLinkComponent
Anti-SOX13 Rabbit Polyclonal Antibody
Anti-SOX13 Rabbit Polyclonal Antibody
Catalog # 102201-576
Supplier:  Novus Biologicals
Anti-SOX13 Rabbit Polyclonal Antibody
Catalog # 102201-576
Supplier:  Novus Biologicals
Supplier Number:  NBP1-71851

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    SRY (sex determining region Y)-box 13
  • Antigen Symbol:
    SOX13
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    9580
  • Antigen Synonyms:
    MGC117216|transcription factor SOX-13|Sox-13|Type 1 diabetes autoantigen ICA12|SRY (sex determining region Y)-box 13ICA12islet cell 12|SRY-related HMG-box gene 13|Islet cell antigen 12|SRY-box 13
  • Storage Buffer:
    Tris-Citrate/Phosphate (pH 7.0 - 8.0)
  • Storage Temperature:
    Store at 4C. Do not freeze.
  • Concentration:
    1 mg/ml
  • Immunogen:
    The immunogen for this product maps to a region between residue 572 and 622 of human Sex Determining Region Y-box 13 using the numbering given in entry NP_005677.2 (GeneID 9580).
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102201-576
  • Supplier no.:
    NBP1-71851

Specifications

About this item

The SOX13 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOX13. This antibody reacts with human. The SOX13 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: SOX13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human