Order Entry
United States
ContactUsLinkComponent
 
Anti-SLC6A5 Rabbit Polyclonal Antibody
  102193-002
Anti-SLC6A5 Rabbit Polyclonal Antibody
  102193-002
 :  NBP1-59657

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Solute Carrier Family 6 (neurotransmitter transporter) Member 5
  • Antigen Symbol:
    GlyT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    9152
  • Antigen Synonyms:
    sodium- and chloride-dependent glycine transporter 2|GLYT-2|GLYT2GlyT-2|NET1|glycine)|member 5|Solute carrier family 6 member 5|GlyT2|solute carrier family 6 (neurotransmitter transporter
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptides corresponding to SLC6A5(solute carrier family 6 (neurotransmitter transporter, glycine), member 5) The peptide sequence was selected from the middle region of SLC6A5. Peptide sequence LIVTCTNSATSIFAGFVIFSVIGFMANERKVNIENV
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102193-002
  • Supplier no.:
    NBP1-59657

 

 

Rabbit Polyclonal SLC6A5 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: SLC6A5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human