Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 22 Member A3
- Antigen Symbol:SLC22A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:6581
- Antigen Synonyms:OCT3EMT organic cation transporter 3|solute carrier family 22 (extraneuronal monoamine transporter)|member 3|solute carrier family 22 member 3|EMTEMTH|Organic cation transporter 3|Extraneuronal monoamine transporter
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to SLC22A3(solute carrier family 22 (extraneuronal monoamine transporter), member 3) The peptide sequence was selected from the N terminal of SLC22A3. Peptide sequence PDHYWCRGPSAAALAERCGWSPEEEWNRTAPASRGPEP
- Purification:Immunogen affinity purified
- Cat. No.:102198-944
- Supplier no.:NBP1-69368
Specifications
About this item
The SLC22A3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC22A3. This antibody reacts with human. The SLC22A3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: SLC22A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human