Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 20 Member A1
- Antigen Symbol:SLC20A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:6574
- Antigen Synonyms:Leukemia virus receptor 1 homolog|GLVR1DKFZp686J2397|FLJ41426|Gibbon ape leukemia virus receptor 1|sodium-dependent phosphate transporter 1|Solute carrier family 20 member 1|member 1|Glvr-1|solute carrier family 20 (phosphate transporter)|Phosphate transporter 1|PiT-1PIT1
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.48 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SLC20A1. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102165-688
- Supplier no.:NBP1-32252
Specifications
About this item
The SLC20A1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC20A1. This antibody reacts with human, mouse. The SLC20A1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: SLC20A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse