Order Entry
United States
ContactUsLinkComponent
 
Anti-SLC1A5 Rabbit Polyclonal Antibody
  102173-904
Anti-SLC1A5 Rabbit Polyclonal Antibody
  102173-904
 :  NBP1-44240
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Solute Carrier Family 1 Member A5
  • Antigen Symbol:
    SLC1A5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    0.05 ml
  • Environmentally Preferable:
  • Gene ID:
    6510
  • Antigen Synonyms:
    ASCT2M7VS1|neutral amino acid transporter B(0)|AAAT|RD114 virus receptor|solute carrier family 1 (neutral amino acid transporter)|neutral amino acid transporter B|RD114/simian type D retrovirus receptor|RDR|R16|RDRCFLJ31068|ATB(0)|member 5|M7V1ATBO|Baboon M7 virus receptor|Solute carrier family 1 member 5|Sodium-dependent neutral amino acid transporter type 2
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1.0 mg/ml
  • Immunogen:
    Synthetic 15 amino acid peptide from C-Terminus of human SLC1A5.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102173-904
  • Supplier no.:
    NBP1-44240

 

 

The SLC1A5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC1A5. This antibody reacts with human. The SLC1A5 Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SLC1A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human