Order Entry
United States
ContactUsLinkComponent
Anti-SLC13A3/NaDC3 Rabbit Polyclonal Antibody
Anti-SLC13A3/NaDC3 Rabbit Polyclonal Antibody
Catalog # 102193-700
Supplier:  Novus Biologicals
Anti-SLC13A3/NaDC3 Rabbit Polyclonal Antibody
Catalog # 102193-700
Supplier:  Novus Biologicals
Supplier Number:  NBP1-60011

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Solute Carrier Family 13 Member A3
  • Antigen Symbol:
    SLC13A3/NaDC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100μl
  • Gene ID:
    64849
  • Antigen Synonyms:
    Sodium-dependent high-affinity dicarboxylate transporter 2|NaDC-3|member 3|sodium-dependent high affinity dicarboxylate transporter 3|solute carrier family 13 (sodium-dependent dicarboxylate transporter)|solute carrier family 13 member 3|hNaDC3|Na(+)/dicarboxylate cotransporter 3|NADC3SDCT2naDC-3
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptides corresponding to SLC13A3(solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3) The peptide sequence was selected from the middle region of SLC13A3 (NP_001011554). Peptide sequence GLSFRGWRKNKSEIRTNAEDRARAVIRE
  • Purification:
    Protein A purified
  • Cat. No.:
    102193-700
  • Supplier no.:
    NBP1-60011

Specifications

About this item

The SLC13A3 / NaDC3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SLC13A3 / NaDC3. This antibody reacts with human. The SLC13A3 / NaDC3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml

Type: Primary
Antigen: SLC13A3/NaDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human