About this item
The SIRT6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SIRT6. This antibody reacts with human. The SIRT6 Antibody has been validated for the following applications: Western Blot, Simple Western.
Type: Primary
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102107-650
- 102107-636
- Antigen Symbol:
- SIRT6
- SIRT6
- Antigen Name:
- Sirtuin 6
- Sirtuin 6
- Antigen Synonyms:
- S. cerevisiae|sirtuin (silent mating type information regulation 2|sirtuin 6|SIR2L6sir2-related protein type 6|SIR2-like protein 6|EC 3.5.1.-|sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae)|sirtuin type 6|homolog) 6|NAD-dependent deacetylase sirtuin-6
- S. cerevisiae|sirtuin (silent mating type information regulation 2|sirtuin 6|SIR2L6sir2-related protein type 6|SIR2-like protein 6|EC 3.5.1.-|sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae)|sirtuin type 6|homolog) 6|NAD-dependent deacetylase sirtuin-6
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 51548
- 51548
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to an internal region of the human SIRT6 protein (within residues 250-350). [Swiss-Prot Q8N6T7]
- A synthetic peptide made to an internal region of the human SIRT6 protein (within residues 250-350). [Swiss-Prot Q8N6T7]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:
- Store at 4C. Do not freeze.
- Store at 4C. Do not freeze.
- Tested Applications:
- Simple Western
- Simple Western