Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:SH3-Domain Binding Protein 2
- Antigen Symbol:SH3BP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Form:Lyophilized
- Gene ID:6452
- Antigen Synonyms:SH3-domain binding protein 2|FLJ54978,3BP-2,3BP2|Abl-SH3 binding protein 2|CRPM|TNFAIP3 interacting protein 2|CRBM|RES4-23|SH3 domain-binding protein 2|Cherubism|FLJ42079
- Modification:p Ser427
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic phospho peptide: SFRSFSpFEKPRQ, corresponding to amino acids 422-433 of Human SH3BP2.
- Purification:Immunogen affinity purified
- Cat. No.:102134-110
- Supplier no.:NB600-492
Specifications
About this item
The SH3BP2 [p Ser427] Antibody from Novus Biologicals is a rabbit polyclonal antibody to SH3BP2. This antibody reacts with human, mouse, rat. The SH3BP2 [p Ser427] Antibody has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.
Type: Primary
Antigen: SH3BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat