Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:SET nuclear oncogene
- Antigen Symbol:SET
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:6418
- Antigen Synonyms:SET nuclear oncogene|I2PP2A|chromatin remodelling factor|TAF-IBETA|2PP2A|SET translocation (myeloid leukemia-associated)|protein phosphatase type 2A inhibitor|Inhibitor of granzyme A-activated DNase|PHAPIITAF-I|protein SET|HLA-DR-associated protein II|Template-activating factor I|I-2PP2A|Phosphatase 2A inhibitor I2PP2A|inhibitor-2 of protein phosphatase-2A|Template-Activating Factor-I|IGAAD|IPP2A2
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.43 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SET. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102167-358
- Supplier no.:NBP1-33713
The SET Antibody from Novus Biologicals is a rabbit polyclonal antibody to SET. This antibody reacts with human, mouse, rat. The SET Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
- Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500
Type: Primary
Antigen: SET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat