Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:RNA Binding Motif Protein 28
- Antigen Symbol:RBM28
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:55131
- Antigen Synonyms:RNA binding motif protein 28|RNA-binding motif protein 28,2810480G15Rik|RNA-binding protein 28|FLJ10377
- Storage Buffer:PBS & 2% Sucrose.
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to RBM28 (RNA binding motif protein 28) The peptide sequence was selected from the C terminal of RBM28. Peptide sequence QTKAEVEQVELPDGKKRRKVLALPSHRGPKIRLRDKGKVKPVHPKKPKPQ.
- Purification:Immunogen affinity purified
- Cat. No.:102189-352
- Supplier no.:NBP1-57228
The RBM28 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RBM28. This antibody reacts with human. The RBM28 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: RBM28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human