Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:6-pyruvoyltetrahydropterin Synthase
- Antigen Symbol:PTS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100μl
- Gene ID:5805
- Antigen Synonyms:EC 4.2.3.12|6-pyruvoyltetrahydropterin synthase|PTPSPTP synthase|6-pyruvoyl tetrahydrobiopterin synthase|FLJ97081
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Full length human PTS Recombinant protein.
- Purification:Immunogen affinity purified
- Cat. No.:102166-602
- Supplier no.:NBP1-32975
Specifications
About this item
The PTS Antibody from Novus Biologicals is a rabbit polyclonal antibody to PTS. This antibody reacts with human, mouse. The PTS Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: PTS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse