Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Protein Kinase D2
- Antigen Symbol:PKC-D2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:25865
- Antigen Synonyms:nPKC-D2|PKD2HSPC187|serine/threonine-protein kinase D2|protein kinase D2|EC 2.7.11|EC 2.7.11.13|DKFZP586E0820
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residues 320 and 400.
- Purification:Immunogen affinity purified
- Cat. No.:102112-776
- Supplier no.:NB100-636
The Protein Kinase D2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Protein Kinase D2. This antibody reacts with human. The Protein Kinase D2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: Protein Kinase D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human