Order Entry
United States
ContactUsLinkComponent
Anti-Occludin Rabbit Polyclonal Antibody
Anti-Occludin Rabbit Polyclonal Antibody
Catalog # 102210-348
Supplier:  Novus Biologicals
Anti-Occludin Rabbit Polyclonal Antibody
Catalog # 102210-348
Supplier:  Novus Biologicals
Supplier Number:  NBP1-77037

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Occludin
  • Antigen Symbol:
    Occludin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    100506658
  • Antigen Synonyms:
    BLCPMG|occludin
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human OCLN. The immunogen is located within the last 50 amino acids of OCLN.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102210-348
  • Supplier no.:
    NBP1-77037

Specifications

About this item

The Occludin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Occludin. This antibody reacts with human, mouse, rat. The Occludin Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 2.5 ?g/ml

Type: Primary
Antigen: Occludin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat