About this item
The NuMA Antibody from Novus Biologicals is a rabbit polyclonal antibody to NuMA. This antibody reacts with human, mouse, rat, mammal, primate. The NuMA Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: NuMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Mammal, Primate
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102131-886
- 102131-888
- Antigen Symbol:
- NuMA
- NuMA
- Antigen Name:
- Nuclear Mitotic Apparatus Protein 1
- Nuclear Mitotic Apparatus Protein 1
- Antigen Synonyms:
- SP-H antigen|structural nuclear protein|centrophilin stabilizes mitotic spindle in mitotic cells|NuMA protein|NUMA|nuclear mitotic apparatus protein 1
- SP-H antigen|structural nuclear protein|centrophilin stabilizes mitotic spindle in mitotic cells|NuMA protein|NUMA|nuclear mitotic apparatus protein 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Primates, Mammals, Human, Rat, Mouse
- Primates, Mammals, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 4926
- 4926
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Full-length recombinant human NuMA expressed in insect Sf9 cells using baculovirus expression system, then purified to near homogeneity prior to immunization.
- Full-length recombinant human NuMA expressed in insect Sf9 cells using baculovirus expression system, then purified to near homogeneity prior to immunization.
- Purification:
- Unpurified
- Unpurified
- Storage Buffer:
- Whole antisera
- Whole antisera
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Tested Applications:
- Simple Western
- Simple Western