Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:NODAL Modulator 1
- Antigen Symbol:NOMO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:23420
- Antigen Synonyms:telomeric copy|Nomo|NOMO3|NODAL modulator 1|pM5 protein 3|pM5 protein
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide directed towards the C terminal of human NOMO1The immunogen for this antibody is NOMO1. Peptide sequence QDIAQGSYIALPLTLLVLLAGYNHDKLIPLLLQLTSRLQGVRALGQAASD.
- Purification:Immunogen affinity purified
- Cat. No.:102214-780
- Supplier no.:NBP1-79566
Specifications
About this item
The NOMO1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NOMO1. This antibody reacts with human. The NOMO1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.
Type: Primary
Antigen: NOMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human