Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Nipped-B homolog (Drosophila)
- Antigen Symbol:NIPBL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:25836
- Antigen Synonyms:IDN3-B|FLJ12597|Nipped-B homolog (Drosophila)|FLJ11203|sister chromatid cohesion 2 homolog|SCC2 homolog|nipped-B-like protein|CDLS1|FLJ13354|Scc2|DKFZp434L1319|delangin|FLJ44854|FLJ13648|IDN3CDLS
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1025 and 1075 of human Nipped-B-like using the numbering given in entry NP_597677.2 (GeneID 25836).
- Purification:Immunogen affinity purified
- Cat. No.:102120-766
- Supplier no.:NB100-93320
The NIPBL Antibody from Novus Biologicals is a rabbit polyclonal antibody to NIPBL. This antibody reacts with human. The NIPBL Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: NIPBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human