Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:NIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:90527
- Antigen Synonyms:dual oxidase maturation factor 1 gamma|mol|dual oxidase maturation factor 1 delta|Numb-interacting protein|homolog of Drosophila Numb-interacting protein|Dual oxidase activator 1|NIPdual oxidase maturation factor 1 alpha|dual oxidase maturation factor 1|FLJ32334|NUMBIPdual oxidase maturation factor 1 beta
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:The specific Immunogen is proprietary information. Peptide sequence PEEGGLLSPRYRSMADSPKSQDIPLSEASSTKAYYRPRRLSLVPADVRGL.
- Purification:Immunogen affinity purified
- Cat. No.:102215-414
- Supplier no.:NBP1-79883
Specifications
About this item
The NIP Antibody from Novus Biologicals is a rabbit polyclonal antibody to NIP. This antibody reacts with human. The NIP Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: NIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human