To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Niemann-Pick C1
- Antigen Symbol:NPC1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Primates,Pig,Chinese hamster,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:4864
- Antigen Synonyms:FLJ98532|type C1|NPC|Niemann-Pick disease|NPC1|Niemann-Pick C1 protein
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118]
- Purification:Immunogen affinity purified
- Cat. No.:102131-636
- Supplier no.:NB400-148H
Specifications
About this item
The Niemann-Pick C1 Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to Niemann-Pick C1. This antibody reacts with human, mouse, rat, porcine, chinese hamster, primate. The Niemann-Pick C1 Antibody [HRP] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NPC1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Chinese Hamster, Primate